Skip to Content
Merck
All Photos(1)

Key Documents

754099

Sigma-Aldrich

diF-TES-ADT

99% (HPLC)

Synonym(s):

2,8-Difluoro-5,11-bis(triethylsilylethynyl)anthradithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36F2S2Si2
CAS Number:
Molecular Weight:
602.95
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

99% (HPLC)

loss

0.5 wt. % TGA, 290 °C

mp

193-198 °C

semiconductor properties

P-type (mobility=1 cm2/V·s)

SMILES string

CC[Si](CC)(CC)C#Cc1c2cc3cc(F)sc3cc2c(C#C[Si](CC)(CC)CC)c4cc5cc(F)sc5cc14

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service