Skip to Content
Merck
All Photos(1)

Key Documents

388572

Sigma-Aldrich

Poly(3,3′-diaminobenzophenone-co-3,3′,4,4′-benzophenonetetracarboxylic dianhydride)

Synonym(s):

LARC-TPI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

powder

dielectric constant

3.3, 1 MHz

mp

>500 °C (lit.)

transition temp

Tg 245 °C
Tm 276 °C (DSC, 20°C/min)

InChI

1S/C17H8N2O5.C13H12N2O/c20-13(7-1-3-9-11(5-7)16(23)18-14(9)21)8-2-4-10-12(6-8)17(24)19-15(10)22;14-11-5-1-3-9(7-11)13(16)10-4-2-6-12(15)8-10/h1-6H,(H,18,21,23)(H,19,22,24);1-8H,14-15H2

InChI key

CLNYWOAUAVHHOL-UHFFFAOYSA-N

Application

Adhesive for wide area bonding of metals, ceramics and polyimide films. Molding powder, varnish component and glass fiber cloth prepreg.

Features and Benefits

Can be imidized at a low temperature and processed as a thermoplastic. High chemical resistance to almost all chemicals and organic solvents. Highly thermo-oxidative and chemically stable polyimide developed by NASA. No weight loss up to 400°C

Physical form

Crystalline (30%) linear polyimide powder.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service