Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5478-55

HiTrap® Protein L

Cytiva 17-5478-55, pack of 5 × 5 mL

Synonym(s):

HiTrap Protein L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

recombinant protein L (E. coli), mammalian free

packaging

pack of 5 × 5 mL

manufacturer/tradename

Cytiva 17-5478-55

storage condition

(20% Ehtanol)

parameter

5 bar

column I.D.

16 mm

matrix

rigid, highly cross-linked agarose

avg. part. size

85 μm (d₅₀v is the median particle size of the cumulative volume distribution)

working range

2-10(pH interval where the medium can be operated without significant change in function.)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

Related Categories

General description

HiTrap® Protein L columns are prepacked with Capto L affinity chromatography resin for purification of antibody fragments containing the kappa light chain. This resin combines a rigid, high-flow agarose matrix with the immunoglobulin-binding recombinant protein L ligand, which has a strong affinity to the variable region of antibody kappa light chains.

Features and Benefits

  • Suitable for the capture of a wide range of antibody fragments such as antigen-binding fragments (Fabs), domain antibodies (dAbs), and single-chain variable fragments (scFvs).
  • Enables a platform approach to antibody fragment purification.
  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of resin.
  • Convenient for process development, as well as small-scale purification.
  • HiTrap™ format enables easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA™ system.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service