Skip to Content
Merck
All Photos(1)

Key Documents

NMuMG

94081121, mouse breast, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

NMuMG, 94081121

biological source

mouse breast

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mammary gland, normal epithelial

Cell Line Description

Initiated from the mammary gland tissue of an adult NAMRU mouse and one of a series of epithelial cell lines derived from normal glandular tissue of mice. The cells exhibit many ultrastructural features of secretory epithelium as well as desmosomes or intermediate junctions, microvilli and tonofibrils. Hemicytes or domes develop when dense monolayers are maintained for several weeks without sub-cultivation. The cells produce benign cystadenomas when inoculated into isogenic mice and are negative for type C oncornaviruses.

Application

NMuMG has been used to compare the actions of insulin like growth factor binding protein 5 (IGFBP-5) with transforming growth factor β 1 (TGFβ1) on epithelial cells. It has also been used to measure the influence of phorbol 12-myristate 13-acetate (PMA) and estradiol on bone morphogenetic protein receptor type 2 (BMPR2) gene expression.

Culture Medium

DMEM + 2mM Glutamine + 10ug/ml Insulin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service