HPLC Analysis of Benzene and Deuterated Benzene on Ascentis® Express C18
Materials
분석 컬럼
column
Ascentis Express C18, 15 cm × 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] acetonitrile; [B] water (55:45, A:B)
flow rate
1.0 mL/min
column temp.
50 °C
detector
UV, 254 nm