Merck
TLC Analysis of Water Soluble Vitamins on HPTLC Silica Gel 60 RP-18 WF254s with concentrating zone
TLC Analysis of Water Soluble Vitamins on HPTLC Silica Gel 60 RP-18 WF254s with concentrating zone application for TLC
Materials
부속물
제품 번호
설명
가격
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Rectangular TLC developing tanks, complete
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Latch-lid TLC developing chambers
For use with 10 cm × 10 cm plates
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid TLC developing chambers
For use with 20 cm × 20 cm plates
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
CONDITIONS
plate
HPTLC pre-coated plate RP-18 F 254s with concentrating zone (1.15498.0001)
eluent
[A] acetonitrile; [B] dichloromethane; [C] water; (90:10:10, A:B:C) containing 0.1 M lithium chloride
migration distance
7 cm
chamber
normal chamber without chamber saturation
sample
normal chamber without chamber saturation
detector
in-situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm