뒤로
Grids for transmission electron microscopy
Products
배송
오늘 출고 (7)
직경(mm)
포장 단위
제품 필터
Showing 1-19 of 19
비교
제품 번호
설명
가격
G4901
Grids for transmission electron microscopy,
grid size 300 mesh × 83 μm pitch, copper
확장
가격 보기
G4776
Grids for transmission electron microscopy,
grid size 200 mesh × 125 μm pitch, copper
확장
가격 보기
G5026
Grids for transmission electron microscopy,
grid size 400 mesh × 62 μm pitch, copper
확장
가격 보기
G1403
Grids for transmission electron microscopy,
grid size 100 mesh × 250 μm pitch, copper
확장
가격 보기
G2153
Grids for transmission electron microscopy,
grid size 1000 mesh × 25 μm pitch, copper
확장
가격 보기
G5651
Grids for transmission electron microscopy,
grid size 300 mesh × 83 μm pitch, nickel
확장
가격 보기
G1653
Grids for transmission electron microscopy,
grid size 150 mesh × 165 μm pitch, copper
확장
가격 보기
G5776
Grids for transmission electron microscopy,
grid size 400 mesh × 62 μm pitch, nickel
확장
가격 보기
G6151
Grids for transmission electron microscopy,
grid size 200 mesh × 125 μm pitch, copper, hole W 115 μm, open area size 84%
확장
가격 보기
G5151
Grids for transmission electron microscopy,
grid size 200 mesh × 125 μm pitch, gold
확장
가격 보기
G1528
Grids for transmission electron microscopy,
grid size 100 mesh × 250 μm pitch, nickel
확장
가격 보기
G2278
Grids for transmission electron microscopy,
grid size 1000 mesh × 25 μm pitch, nickel
확장
가격 보기
G5526
Grids for transmission electron microscopy,
grid size 200 mesh × 125 μm pitch, nickel
확장
가격 보기
G5901
Grids for transmission electron microscopy,
grid size 200 mesh × 125 μm pitch, copper, hole W 95 μm, open area size 55%
확장
가격 보기
G5276
Grids for transmission electron microscopy,
grid size 300 mesh × 83 μm pitch, gold
확장
가격 보기
G1903
Grids for transmission electron microscopy,
grid size 600 mesh × 42 μm pitch, copper
확장
가격 보기
G5401
Grids for transmission electron microscopy,
grid size 400 mesh × 62 μm pitch, gold
확장
가격 보기
G2028
Grids for transmission electron microscopy,
grid size 600 mesh × 42 μm pitch, nickel
확장
가격 보기
G1778
Grids for transmission electron microscopy,
grid size 150 mesh × 165 μm pitch, nickel