주요 문서
K5141-U
Omnifit® EZ Benchmark Columns with 2 Fixed Endpieces
I.D. × L 15 mm × 400 mm
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩743,183
₩743,183
예상 입고일2025년 8월 08일세부사항
벌크 견적 요청
크기 선택
보기 변경
1 EA
₩743,183
About This Item
UNSPSC 코드:
23151816
NACRES:
SB.52
₩743,183
예상 입고일2025년 8월 08일세부사항
벌크 견적 요청
제조업체/상표
Omnifit 006EZ-15-40-FF
Quality Level
기술
LPLC: suitable
I.D. × 길이
15 mm × 400 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Omnifit® Labware complete columns are supplied assembled and ready for use, with tubing and fittings to connect the column into an existing system. Two configurations are available:
  • Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
  • One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
법적 정보
Omnifit is a registered trademark of Diba Industries
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)