주요 문서
20857
Tubing Bender
Three-Size for 1/4in. (6 mm, 3/16in. (4 mm) & 1/8in. tube
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
일반 설명
The heavy duty benders easily bend aluminum, copper, or stainless steel tubing up to 180°, without kinking or splitting the tubing.
The three-in-one tool (20857) bends 1/8 in., 3/16 in., or 1/4 in. (3, 4, or 6 mm) tubing of standard wall thickness.
20422-U has a bend radius of 3/8 inch from center.
20424-U has a bend radius of 9/16 inch from center.
20857 (three-in-one tool) has a bend radius of 14.2 mm from center (pictured above).
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)