주요 문서
20668
Supelco
Thermogreen® LB-1 Septa, cylindrical
septum type cylindrical with half-hole
, pkg of 100 ea
로그인조직 및 계약 가격 보기
크기 선택
100 EA
₩231,928
₩231,928
예상 입고일2025년 4월 07일세부사항
벌크 견적 요청
크기 선택
보기 변경
100 EA
₩231,928
About This Item
UNSPSC 코드:
41115717
₩231,928
예상 입고일2025년 4월 07일세부사항
벌크 견적 요청
포장
pkg of 100 ea
직경 × 길이
~6 mm × 9 mm
응용 분야
environmental
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
For use in glass gas sampling bulbs, and purge & trap glassware.
법적 정보
Thermogreen is a registered trademark of Merck KGaA, Darmstadt, Germany
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Elefteria Psillakis et al.
Chemosphere, 54(7), 849-857 (2003-11-26)
The sonochemical degradation of aqueous solutions containing low concentrations of six phthalate esters at an ultrasonic frequency of 80 kHz has been investigated. Ultrasonic treatment was found capable of removing the four higher molecular mass phthalates (di-n-butyl phthalate, butylbenzyl phthalate
Elefteria Psillakis et al.
Journal of chromatography. A, 999(1-2), 145-153 (2003-07-30)
A simple and efficient liquid-phase microextraction (LPME) technique using a hollow-fibre membrane, in conjunction with gas chromatography-mass spectrometry has been developed for the extraction and analysis of six phthalate esters in water samples. Parameters such as extraction solvent, agitation of
Solid-phase microextraction (SPME) for rapid field sampling and analysis by gas chromatography-mass spectrometry (GC-MS).
Hook, Gary L., et al.
TrAC, Trends in Analytical Chemistry, 21, 534-543 (2002)
R.L. Grob, M.A. Kaiser
Environmental Problem Solving Using Gas and Liquid Chromatography, 166-166 (2000)
Robert L. Grob, PhD, Eugene F. Barry, PhD
Modern Practice of Gas Chromatography, 471-472 (2004)