주요 문서
2047825
Supelco
Inlet Liner, Split/Splitless Type, Single Taper Design (wool packed)
pkg of 25 ea
로그인조직 및 계약 가격 보기
크기 선택
25 PKG
₩916,342
₩916,342
구입 가능 여부는 고객센터에 문의하십시오.
벌크 견적 요청
크기 선택
보기 변경
25 PKG
₩916,342
About This Item
UNSPSC 코드:
41121800
₩916,342
구입 가능 여부는 고객센터에 문의하십시오.
벌크 견적 요청
포장
pkg of 25 ea
길이 × O.D. × I.D.
78.5 mm × 6.5 mm × 4.0 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)
The liner is the second most important part of the inlet. It is the centrepiece of the inlet system, basically a glass tube between the septum and the column, where the sample is evaporated and brought into the gas phase. Split/splitless inlet allows fast sample introduction and vaporization which is highly advantageous for small sample sizes. Though the injection modes for both split and splitless are carried out on the same inlet, there are certain differences like a splitless injection involves the complete introduction of the sample into the column whereas in a split injection, the sample is divided in the inlet as a result only part of the sample is introduced in the column.[1]
애플리케이션
Split/splitless inlet equipped with a flip-top sealing system in a thermal desorption unit maybe used to perform thermal desorption (TD) for the determination of pyrethroids in water samples.[2]
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Els Van Hoeck et al.
Journal of chromatography. A, 1157(1-2), 1-9 (2007-05-08)
Stir bar sorptive extraction (SBSE) was applied to the enrichment of pyrethroids in water samples. Analysis was done by retention time-locked GC/MS in selected ion-monitoring (SIM) mode using trans-permethrin-d(6) as internal standard. Thermal desorption (TD) was performed in a classical
Fire Debris Analysis.
Stauffer E
Science, 249-252 (2007)
문서
Analysis of carbonate solvents in lithium-ion batteries like ethylene carbonate, dimethyl carbonate, diethyl carbonate, and more using SPB®-624 column.