주요 문서
Z709948
Sartorius pipette tips
volume range 5-350 μL, standard, refill, non-sterile
로그인조직 및 계약 가격 보기
크기 선택
960 EA
₩102,515
₩102,515
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
960 EA
₩102,515
About This Item
UNSPSC 코드:
41121600
NACRES:
NB.01
₩102,515
구입 가능 여부는 고객센터에 문의하십시오.
material
colorless
무균
non-sterile
특징
DNase free
RNase free
barrier: no
endotoxin free
포장
pack of 96 ea (Racked)
제조업체/상표
Sartorius 790352
최대 용적
350 μL
용적 범위
5-350 μL
용적
5—350 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
기타 정보
Browse all pipette tips
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)