주요 문서
Z327891
BRAND® Transferpette® pipette, fixed volume, single channel
volume 200 μL, accuracy: 0.6%, precision: 0.2%
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
가격 및 재고 정보를 현재 이용할 수 없음
설명
corrosion resistant piston
supplied with performance certificate and calibration certificate.
특징
electronic: no
recommended tips 50-1000 μL & 50-1250 μL
single-channel
제조업체/상표
BRAND
BrandTech 704146
부피 측정 범위
200 μL, accuracy: 0.6%, precision: 0.2%
용적
200 μL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Transferpette pipette is designed to the shape of the human hand for maximum comfort. The side mounted pipetting key reduces hand fatigue, especially when performing serial pipetting operations. The textured housing of the hand grip ensures a firm grip and is UV resistant.
Changing volume is easy without snagging gloves. Detents prevent inadvertent volume changes. Easy to calibration tool supplied. The slim shaft (autoclavable at 121°C) makes it possible to pipette in the narrowest vessels without removing the tip ejector. The taper is designed to fit PLASTIBRAND and most other leading manufacturers tips.
CE-marked according to IVD-Directive 98/79 CE.
* Filter tips
기타 정보
BR704146 (India)
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)