주요 문서
TMO3663-0250
Nalgene® graduated cylinders
PMP, volume 250 mL, case of 12 ea
동의어(들):
PMP cylinder, graduated PMP cylinder
로그인조직 및 계약 가격 보기
크기 선택
12 EACH
₩925,579
₩925,579
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
12 EACH
₩925,579
About This Item
UNSPSC 코드:
41121805
NACRES:
NB.43
₩925,579
구입 가능 여부는 고객센터에 문의하십시오.
material
PMP
포장
case of 12 ea
제조업체/상표
Nalgene 3663-0250
용적
250 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Non-wetting, they deliver all they contain (calibrated TC/TD at 20°C). Meet Class B accuracy requirements of ASTM E1272 "Cylinder, Graduated, Laboratory, Glass" and all requirements of ISO Standard 6706 "Plastic Laboratory Ware-Graduated Measuring Cylinders". No meniscus to confuse readings. Blue, molded, PP base resists tipping. Molded-in graduations with additional major graduation lines on the back of cylinder for easier, more accurate fluid level readings. Generous pour spout. Can be autoclaved, but accuracy will be affected.
법적 정보
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)