주요 문서
HS120649
Reagent Reservoir
capacity 100 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
로그인조직 및 계약 가격 보기
크기 선택
100 EACH
₩156,006
₩156,006
예상 입고일2025년 6월 09일세부사항
크기 선택
보기 변경
100 EACH
₩156,006
About This Item
UNSPSC 코드:
41121522
NACRES:
NB.01
₩156,006
예상 입고일2025년 6월 09일세부사항
material
V-bottom bottom
polystyrene
무균
non-sterile
포장
pack of 100 (bulk packed)
제조업체/상표
Heathrow Scientific 120649
용량
100 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
특징 및 장점
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)