주요 문서
GE18-3216-03
Labmate PD-10 Buffer Reservoir
Cytiva 18-3216-03, pack of 1 ea
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41115700
NACRES:
NA.56
가격 및 재고 정보를 현재 이용할 수 없음
무균
non-sterile
특징
wetted part: no
포장
pack of 1 ea
제조업체/상표
Cytiva 18-3216-03
일반 설명
LabMate PD-10 buffer reservoir.
특징 및 장점
  • Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex® G-25 Medium.
  • LabMate Buffer Reservoir makes it easier and more convenient to equilibrate PD-10 columns.
  • Easy to use and equilibrate; apply sample; elute.
법적 정보
Sephadex is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서
This page describes characteristics of Glutathione Sepharose and HiTrap® Benzamidine FF (high sub) media and columns from Cytiva.
Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.
프로토콜
This page shows how to separate IgG antibodies by affinity chromatography using Protein G GraviTrap from Cytiva.
This page shows how to purify histidine-tagged recombinant proteins by gravity flow using His GraviTrap and His GraviTrap Kit from Cytiva.
This page describes efficient column packing and preparation for affinity chromatography of GST-tagged proteins using Cytiva products.
Spectrophotometry can be used to estimate DNA or RNA concentration and to analyze the purity of the preparation.