주요 문서
BMSC1005SC5OB
Sigma-Aldrich
Microtube Centrifuge Tubes
capacity 5 mL, black tube, sterile, cap (screw, attached), pack of 500 ea
로그인조직 및 계약 가격 보기
크기 선택
500 EACH
₩224,011
₩224,011
예상 입고일2025년 4월 28일세부사항
크기 선택
보기 변경
500 EACH
₩224,011
About This Item
UNSPSC 코드:
41121703
NACRES:
NB.11
₩224,011
예상 입고일2025년 4월 28일세부사항
material
black tube
cap (screw, attached)
conical bottom
polypropylene tube
무균
sterile
특징
DNase free
RNase free
graduations
포장
pack of 500 ea
제조업체/상표
Benchmark Scientific C1005-SC5-OB
용량
5 mL
튜브 직경
16 mm
적합성
suitable for (centrifugation)
suitable for PCR
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
5mL MacroTubes take all of the conveniences of standard 15mL conical tubes and scales them down for space efficiency and decreased natural resource consumption. Useful from -86°C to 110°C, these tubes are a valuable resource for boiling and for sample preservation at very low temperatures. Choose from gamma sterilized racked packaging with attached caps or non-sterile bulk packaging with separate caps. The foam racks are similar to the packaging normally associated with 15mL tubes (upright with caps attached), but they require about 60% less storage space.
애플리케이션
Centrifugation, PCR
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)