주요 문서
11-0320
Sigma-Aldrich
Florisil®
CP, 100-200 mesh
동의어(들):
Activated magnesium silicate, Magnesium silicate
로그인조직 및 계약 가격 보기
About This Item
실험식(Hill 표기법):
MgO3Si
CAS Number:
Molecular Weight:
100.39
EC Number:
MDL number:
UNSPSC 코드:
12000000
PubChem Substance ID:
가격 및 재고 정보를 현재 이용할 수 없음
Grade
CP
양식
powder
재고 정보
available only in Japan
표면적
289 m2/g
입자 크기
100-200 mesh
SMILES string
[Mg++].[O-][Si]([O-])=O
InChI
1S/Mg.O3Si/c;1-4(2)3/q+2;-2
InChI key
FKHIFSZMMVMEQY-UHFFFAOYSA-N
유사한 제품을 찾으십니까? 방문 제품 비교 안내
법적 정보
Florisil is a registered trademark of U.S. Silica Company
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
개인 보호 장비
Eyeshields, Gloves, type N95 (US)
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Alain Hildebrandt et al.
Archives of environmental contamination and toxicology, 57(2), 247-255 (2008-12-05)
This study was aimed to assess the presence and fate of 22 organochlorinated pesticides (OCHs) and their degradation products and 16 Environmental Protection Agency-priority polycyclic aromatic hydrocarbons (PAHs) in soils of the Ebro River basin (NE Spain) during a 3-year
U Avsar et al.
Cellular and molecular biology (Noisy-le-Grand, France), 59 Suppl, OL1835-OL1841 (2013-02-13)
This study evaluated the protective effect of Panax Ginseng (PG) on bone metabolism in an experimental ovariectomy (OVX) model of osteoporosis in which inflammation was induced by subcutaneous magnesium silicate. The groups were: sham control (Group1, SH), sham+inflammation (Group2, SHinf)
Ting Zhou et al.
Analytical chemistry, 84(13), 5816-5822 (2012-06-13)
Microwave accelerated selective Soxhlet extraction (MA-SSE), a novel selective extraction technique, was investigated in this study. A Soxhlet extraction system containing a glass filter was designed as an extractor. During the procedure of MA-SSE, both the target analytes and the
N García-Flor et al.
Environmental technology, 29(12), 1275-1283 (2009-01-20)
A reliable analytical procedure was developed to determine trace levels of organochlorine compounds in neuston samples. Freeze-dried samples were extracted by sonication (3 x 15 ml cyclohexane, 15 min) followed by gel permeation chromatography (GPC) and gas chromatography electron capture
Youtao Xie et al.
Acta biomaterialia, 5(6), 2331-2337 (2009-04-14)
In this paper, chemically synthesized Mg(2)SiO(4) (MS) powder was plasma-sprayed onto a titanium alloy substrate to evaluate its application potentials in biomedicine. The phase composition and surface morphology of the MS coating were analyzed. Results showed that the MS coating