YY3029358
1 of 10
Millipore
YY2214253
O-ring, 2-251, PTFE
Sigma-Aldrich
74146
Sterile disks
XX6700059
O-ring, fluoroelastomer A
70191
Mueller Hinton Agar
YY3014234
Support Screen for 142 mm filter holder, PTFE-coated
S6141
Wheaton staining dishes
XX6700P10
Dispensing Pressure Vessel, 10 L
XX1100000
Pressure Vessel, 600 mL
YY3014236
Filter Holder 142 mm, stainless steel
XX6700P01
Dispensing Pressure Vessel, 1 gal
for use with 293-mm Stainless Steel Sanitary Filter Holder
100
제품 번호
로트/배치 번호