주요 문서
TANKSDS2UV
Milli-Q
Loop UV Lamp Assembly 254 nm
Maintains consistent purity of stored water
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104200
NACRES:
JA.13
가격 및 재고 정보를 현재 이용할 수 없음
포장
pkg of 1 unit
파라미터
254 nm (wavelength)
호환성
for use with SDS
일반 설명
This water purification accessory is designed for use with SDS 500 storage tanks. Pure water in the reservoir is distributed to multiple locations through the loop. This 254 nm UV lamp limits bacterial growth in the loop, thereby protecting the quality of pure water.
애플리케이션
This accessory prevents bacterial proliferation and biofilm buildup in the distribution loop.
면책조항
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.