주요 문서
TANKLKXL1
Milli-Q
Water sensor
For high volume water purification systems
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104200
NACRES:
JA.13
가격 및 재고 정보를 현재 이용할 수 없음
포장
pkg of 1 unit
호환성
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
This water sensor is a safety accessory which detects water spillage on the floor and automatically closes the system′s inlet solenoid valve to shut down the feed water access from the system or main water supply. The water sensor is typically placed on the floor or benchtop, where most convenient for the lab.
애플리케이션
This water sensor accessory for high volume water purification system prevents water damage to the lab, in an unlikely event of a leak.
특징 및 장점
  • Highly reduces the risk of flooding.
  • Protects instruments and lab surroundings from water damage.
법적 정보
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany