주요 문서
SQPAKT0CM1
Milli-Q
SQPAK TOC Quanta Cartridge
greener alternative
Produces ultrapure (Type 1) water with TOC ≤ 5 ppb. Use with the ultrapure dispensing module of Milli-Q® SQ 2Series systems.
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104212
eCl@ss:
33050190
NACRES:
NB.35
가격 및 재고 정보를 현재 이용할 수 없음
Quality Level
포장
pkg of 1 unit
환경친화적 대안 제품 특성
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
호환성
for use with Milli-Q® SQ 2Series
환경친화적 대안 카테고리
일반 설명
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The SQPAK TOC Quanta cartridge produces ultrapure (Type 1) water at 18.2 MΩ·cm resistivity and TOC ≤ 5 ppb.
The cartridge contains IQnano ion-exchange media to remove ions down to trace levels and high-grade synthetic activated carbon that targets traces of organic contaminants.
For use on the ultrapure dispensing module of Milli-Q® SQ 2Series configurations, including Milli-Q® SQ 200/200P and SQ 240C/240CV systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
특징 및 장점
  • Easy replacement thanks to the Twist & Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
  • Ultrapure label and blue color of the SQPAK Quanta cartridge are repeated on the production station interface, to inform about its water purification purpose and guide replacement  
법적 정보
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany
문서 라이브러리 방문
프로토콜
Milli-Q® SQ 2Series 실험용 정제수 시스템에 대한 지침 영상을 보고 사용자 설명서를 다운로드하십시오. 즉시 설치하여 사용할 수 있습니다. 시스템, 액세서리, 카트리지 및 서비스를 온라인으로 구매하십시오.
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
관련 콘텐츠
초순수에 대한 새로운 방식에 대해 알아보세요. 이제 플러그가 있는 모든 실험실 벤치에서 18.2 MΩ∙cm 초순수를 신선하게 채수할 수 있습니다. 견고한 품질. 간편함. 컴팩트. 확장성. 친환경 대체 제품. 온라인 구매.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.