167521
1.67521
1 of 10
Supelco
1.01114
Withdrawal system
1.04936
Potassium chloride
Sigma-Aldrich
L8662
L-Lysine monohydrochloride
1.02400
Potassium hydrogen phthalate
22668-U
Swagelok® Fittings Kit
50652-U
Ascentis® Express 90 Å PCS-C18, 2.7 µm HPLC Column
47927
CEN PCB Congener Mix-1
SAFC
1.06522
Sodium-(S)-lactate-solution
Millipore
VT000133
Escherichia coli WDCM 00013 Vitroids™
1.00334
Hydrofluoric acid 48%
no temp limit
제품 번호
로트/배치 번호