material
black polystyrene
특징
Irradiated with lid, skirtless
포장
pkg of 5 ea
제조업체/상표
Whatman 7716-2370
Whatman Article No. 28420921 (US reference)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Whatman glass bottom microplates are designed for high-sensitivity detection including fluorescent and luminescent detection and scintillation counting, where extremely low backgrounds with no cross talk are needed. Glass bottom microplates have excellent uniformity in flatness and thickness (0.175 mm glass thickness) to provide optically clear as well as optically flat surfaces. This ensures confluence and planarity for confocal imaging and detection techniques.
They are suitable for FRET and GFP. The skirtless glass bottom plate allows the bottom of the plate to be positioned very close to microscope objectives. This is the plate of choice for Zeiss Confocal Microscopes. The glass bottom microplates are available in clear and black in a 96 well format.
Features and Benefits
Applications
They are suitable for FRET and GFP. The skirtless glass bottom plate allows the bottom of the plate to be positioned very close to microscope objectives. This is the plate of choice for Zeiss Confocal Microscopes. The glass bottom microplates are available in clear and black in a 96 well format.
Features and Benefits
- Superior optical clarity
- Optics using single or dual wavelength probe
- Sensitivity
- Absolute flatness
Applications
- Receptor-ligand interaction
- DNA-protein interaction
- Enzyme studies
- Cell based assays
기타 정보
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
법적 정보
Whatman is a registered trademark of Cytiva
가장 최신 버전 중 하나를 선택하세요:
자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..
고객지원팀으로 연락바랍니다.