Z682063
Ace rotary evaporator condenser complete "C" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144, includes 1 L receiving flask and clamp
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩2,900,058
₩2,900,058
구입 가능 여부는 고객센터에 문의하십시오.
모든 사진(1)
크기 선택
보기 변경
1 EA
₩2,900,058
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.43
₩2,900,058
구입 가능 여부는 고객센터에 문의하십시오.
추천 제품
일반 설명
The "C" condenser assembly consists of the main glass outer trap body with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks.
Features standard top and side flange that connects directly to the Büchi® rotary evaporator. The assembly also includes the glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Features standard top and side flange that connects directly to the Büchi® rotary evaporator. The assembly also includes the glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
법적 정보
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
가장 최신 버전 중 하나를 선택하세요:
활성 필터