주요 문서
Z566039
Sigma-Aldrich
Pure-Pac® II dispense coupler kit
1/2in. tube end
로그인조직 및 계약 가격 보기
크기 선택
1 KIT
₩1,787,517
₩1,787,517
출고 가능일2025년 4월 21일세부사항
크기 선택
보기 변경
1 KIT
₩1,787,517
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.01
₩1,787,517
출고 가능일2025년 4월 21일세부사항
일반 설명
The Sigma-Aldrich Pure-Pac II returnable containers require the Micromatic MacroValve coupler for dispensing chemical. The Dispense Head Coupler Kit includes the MacroValve coupler along with the standard components needed to connect to either 1/2in. or 3/8in. diameter transfer lines. All of the hardware in the kit is supplied in a foam-lined plastic case. See Technical Bulletin AL-236 for assembly instructions.
법적 정보
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany
키트 구성품 역시 별도로 이용 가능함
제품 번호
설명
SDS
  • Z104388PTFE sealing tape, W 1/2 in.SDS
  • Adapter for Kilo-Lab® and Pure-Pac® cylinders, ferrule size 1/2 in., NPTM 1/2 in.
  • Z560723Micromatic MacroValve coupler for returnable containersSDS
  • Swagelok® hex reducing coupling, pkg of 1 ea
  • Swagelok® street elbow, pkg of 1 ea
  • Z565547Stainless steel hose connector, 1/4in. male NPT, 1/4in. hose I.D., pkg of 1 eaSDS
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)