문서
Z507318
Supelco
Michel-Miller Swagelok® adapter
PTFE, NPTF 1/16 in., Ace-Thred for #25
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12000000
NACRES:
SB.52
material
PTFE
제조업체/상표
Ace Glass 584448
기술
LPLC: suitable
NPTF
1/16 in.
피팅
Ace-Thred for #25
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
법적 정보
Swagelok is a registered trademark of Swagelok Company