주요 문서
Z406511
Stankovic transfer adapter
joint: ST/NS 24/29, thread size 13-425
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩109,011
₩109,011
예상 입고일2025년 6월 05일세부사항
크기 선택
보기 변경
1 EA
₩109,011
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.35
₩109,011
예상 입고일2025년 6월 05일세부사항
나사산 크기
13-425
조인트
joint: ST/NS24/29
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
These unique adapters greatly simplify the transfer of solids contained in round-bottom flasks into vials. Precision machined, chemically inert PTFE adapters will not seize in the joint. A wide range of thread sizes are available to accommodate most sample vials including scintillation vials (22 mm threads).
  • Transfers samples without exposure to air or moisture
  • Reduces sample losses due to air currents and static charge that can normally cause light solids to float or blow away when transferred open to the air
  • Excellent for transferring fluffy lyophilized samples, especially peptides
  • Transfers any freely flowing solid and eliminates weighing paper or other intermediate devices
Easy to Use:
Screw sample vial into bottle thread at top of adapter. Insert other end of adapter into flask joint. Invert assembly and gently tap vial on a soft surface to transfer solids from flask into sample vial.
기타 정보
Care must be used when tapping vial to prevent accidental breakage. Tapping on a cork ring or other soft surface is recommended.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)