주요 문서
Z341665
3M 6000/7000 series filters
ABE1 Organic Vapour, inorganic and gas filter, pkg of 2 ea
로그인조직 및 계약 가격 보기
크기 선택
1 PKG
₩39,382
₩39,382
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
1 PKG
₩39,382
About This Item
UNSPSC 코드:
46182002
NACRES:
NB.35
₩39,382
구입 가능 여부는 고객센터에 문의하십시오.
설명
Protects against organic vapours with b.p. >65°C, inorganic and acid gases up to 10 × OEL* or 1000ppm*, EN141
포장
pkg of 2 ea
제조업체/상표
3M 6057
재고 정보
not available in USA and Canada
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Gas and vapour filters
Two filters required per respirator.
특징 및 장점
These filters utilise 3M′s advanced filter technology which minimises breathing resistance for greater comfort to the user. Select gas and vapour filters, particulate filters according to protection required. All filters fit both 6000 and 7000 series respirators by bayonet connection. Two filters required per respirator.
경고
All these filters should only be used for protection against hazards with good warning properties, e.g. smell or taste, and providing the concentrations of hazardous substances are not immediately dangerous to life or health (IDLH). *Where concentrations are given in terms of the OEL or ppm, the filter should only be used for protection against concentrations below the lower value.
법적 정보
3M is a trademark of 3M Company
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)