주요 문서
Z277827
Sigma-Aldrich
HGS Space-filling molecular models
Researcher set
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
60104506
NACRES:
NB.17
제조업체/상표
HGS 2001
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Designed for building biomolecules and complexed molecules such as steroids and peptides. The idea of α-helix or β -sheet in peptide molecules can be visualized. Suited for studying biosynthesis through nucleotides. 383 pieces.
These models combine the idea of both ball-and-stick and space-filling models for visualization of molecular frameworks as well as molecular structures covered with electron clouds. Color-coded model parts are made of flexible polyacetal covered with transparent polyacrylate. Can be used interchangeably with HGS biochemistry molecular models.
Elementary set= 137 pieces
Research set= 383 pieces
Scale: 1 Å = 1.0 cm.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)