์ฝ˜ํ…์ธ ๋กœ ๊ฑด๋„ˆ๋›ฐ๊ธฐ
Merck
๋ชจ๋“  ์‚ฌ์ง„(1)

์ฃผ์š” ๋ฌธ์„œ

S38386

(1-METHOXY-CYCLOPENTYL)-BENZENE

AldrichCPR

๋กœ๊ทธ์ธ์กฐ์ง ๋ฐ ๊ณ„์•ฝ ๊ฐ€๊ฒฉ ๋ณด๊ธฐ

ํฌ๊ธฐ ์„ ํƒ

S38386-100MG
โ‚ฉ47,500

โ‚ฉ47,500


์˜ˆ์ƒ ์ž…๊ณ ์ผ2025๋…„ 4์›” 25์ผ์„ธ๋ถ€์‚ฌํ•ญ



ํฌ๊ธฐ ์„ ํƒ

๋ณด๊ธฐ ๋ณ€๊ฒฝ
S38386-100MG
โ‚ฉ47,500

About This Item

Linear Formula:
C12H16O
CAS Number:
Molecular Weight:
176.261
MDL number:

โ‚ฉ47,500


์˜ˆ์ƒ ์ž…๊ณ ์ผ2025๋…„ 4์›” 25์ผ์„ธ๋ถ€์‚ฌํ•ญ


๊ธฐํƒ€ ์ •๋ณด

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

๊ฐ€์žฅ ์ตœ์‹  ๋ฒ„์ „ ์ค‘ ํ•˜๋‚˜๋ฅผ ์„ ํƒํ•˜์„ธ์š”:

์‹œํ—˜ ์„ฑ์ ์„œ(COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our ๋ฌธ์„œ section.

๋„์›€์ด ํ•„์š”ํ•˜์‹œ๋ฉด ์—ฐ๋ฝํ•˜์„ธ์š”. ๊ณ ๊ฐ ์ง€์› ๋ถ€์„œ

์ด ์ œํ’ˆ์„ ์ด๋ฏธ ๊ฐ€์ง€๊ณ  ๊ณ„์‹ญ๋‹ˆ๊นŒ?

๋ฌธ์„œ ๋ผ์ด๋ธŒ๋Ÿฌ๋ฆฌ์—์„œ ์ตœ๊ทผ์— ๊ตฌ๋งคํ•œ ์ œํ’ˆ์— ๋Œ€ํ•œ ๋ฌธ์„œ๋ฅผ ์ฐพ์•„๋ณด์„ธ์š”.

๋ฌธ์„œ ๋ผ์ด๋ธŒ๋Ÿฌ๋ฆฌ ๋ฐฉ๋ฌธ

์ž์‚ฌ์˜ ๊ณผํ•™์žํŒ€์€ ์ƒ๋ช… ๊ณผํ•™, ์žฌ๋ฃŒ ๊ณผํ•™, ํ™”ํ•™ ํ•ฉ์„ฑ, ํฌ๋กœ๋งˆํ† ๊ทธ๋ž˜ํ”ผ, ๋ถ„์„ ๋ฐ ๊ธฐํƒ€ ๋งŽ์€ ์˜์—ญ์„ ํฌํ•จํ•œ ๋ชจ๋“  ๊ณผํ•™ ๋ถ„์•ผ์— ๊ฒฝํ—˜์ด ์žˆ์Šต๋‹ˆ๋‹ค..

๊ณ ๊ฐ์ง€์›ํŒ€์œผ๋กœ ์—ฐ๋ฝ๋ฐ”๋ž๋‹ˆ๋‹ค.