GF81924161
동의어(들):
FE220330
1 of 10
Sigma-Aldrich
356018
Zinc
349178
Copper
Z645141
IKA® M 20 universal mill
GF41889808
326852
Aluminum
182702
Poly(vinylidene fluoride)
268259
Nickel
267619
GF88259361
Polyethylene Naphthalate
BR708220
Filling tube for Dispensette® S Organic / S Organic
foil
Goodfellow 819-241-61
500 × 500 mm
0.7 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.