문서
DWKW216933
Wheaton wide-mouth bottle
wide-mouth amber soda-lime glass bottle, capacity (1,000 mL), flat PTFE
동의어(들):
W216933
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.17
material
PTFE liner
flat PTFE
polypropylene cap
wide-mouth amber soda-lime glass bottle
특징
with cap
with cap
포장
case of 4 ea
제조업체/상표
WHEATON® W216933
보틀 직경 × 높이
(157 mm) (256 mm)
용량
(1,000 mL)
피팅
89-400
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Wheaton wide-mouth bottle is the largest size available in Type III glass bottles, specifically designed for general sample storage. Its wide-mouth opening facilitates easy filling and pouring, making it convenient for handling various types of samples. Constructed from durable glass, this bottle provides excellent chemical resistance, ensuring the integrity of the stored contents. Ideal for laboratories and industrial applications, it offers a reliable solution for managing bulk samples and liquids efficiently.
특징 및 장점
• Available in larger bottle sizes• Wide mouth / neck• Clear, Type III soda-lime glass• Available with caps attached or without caps• HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification• Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
법적 정보
WHEATON is a registered trademark of Wheaton Industries, Inc.