주요 문서
DWK92812F-25
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (25 mL), class A
로그인조직 및 계약 가격 보기
크기 선택
6 EA
₩473,295
₩473,295
예상 입고일2025년 5월 30일세부사항
크기 선택
보기 변경
6 EA
₩473,295
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
₩473,295
예상 입고일2025년 5월 30일세부사항
material
PTFE stopper
glass flask
특징
class A
포장
case of 6 ea
제조업체/상표
Kimble® 92812F-25
플라스크 용량
(25 mL)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
특징 및 장점
  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)