주요 문서
DWK14396-10000
KIMBLE® KIMAX® Media bottle
without cap, round transparent borosilicate glass bottle, bottle capacity (10,000 mL), non-sterile
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩549,217
₩549,217
예상 입고일2025년 7월 01일세부사항
크기 선택
보기 변경
1 EA
₩549,217
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.43
₩549,217
예상 입고일2025년 7월 01일세부사항
material
round transparent borosilicate glass bottle
무균
non-sterile
특징
without cap
포장
case of 1 ea
제조업체/상표
Kimble® 14396-10000
보틀(bottle) 용량
(10,000 mL)
피팅
GL 45 neck
GL 45
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
특징 및 장점
• Enhanced graduations and marking spot made with chemically resistant white enamel paint• 30 mm ID opening• Autoclavable• Supplied without caps or with linerless GL 45 screw thread caps• Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
법적 정보
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)