주요 문서
CDS024250
Sigma-Aldrich
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazolo[1,5-a]pyrimidine
로그인조직 및 계약 가격 보기
크기 선택
25 MG
₩190,414
₩190,414
예상 입고일2025년 3월 20일세부사항
벌크 견적 요청
크기 선택
보기 변경
25 MG
₩190,414
About This Item
실험식(Hill 표기법):
C12H16BN3O2
Molecular Weight:
245.09
MDL number:
UNSPSC 코드:
12352200
PubChem Substance ID:
₩190,414
예상 입고일2025년 3월 20일세부사항
벌크 견적 요청
설명
AldrichCPR
양식
solid
SMILES string
CC1(OB(C2=C3N=CC=CN3N=C2)OC1(C)C)C
InChI
1S/C12H16BN3O2/c1-11(2)12(3,4)18-13(17-11)9-8-15-16-7-5-6-14-10(9)16/h5-8H,1-4H3
InChI key
ZPGVBVUVOXWXDJ-UHFFFAOYSA-N
기타 정보
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class Code
11 - Combustible Solids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)