주요 문서
CAD6188
1-way threaded end adapter (UTS)
MLL to 5/16-28 standard thread (plated brass)
로그인조직 및 계약 가격 보기
크기 선택
10 EA
₩479,385
₩479,385
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
10 EA
₩479,385
About This Item
UNSPSC 코드:
41121815
NACRES:
NB.03
₩479,385
구입 가능 여부는 고객센터에 문의하십시오.
포장
pack of 10 ea
제조업체/상표
Cadence Science 6188
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
법적 정보
Micro-Mate is a registered trademark of Cadence Science
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)