주요 문서
BR88660
BRAND® stopcock T-bore with nozzles
for tube I.D. 5 mm
동의어(들):
PTFE, Stopcocks
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩125,454
₩125,454
예상 입고일2025년 5월 23일세부사항
크기 선택
보기 변경
1 EA
₩125,454
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.13
₩125,454
예상 입고일2025년 5월 23일세부사항
material
PTFE
포장
pack of 1 ea
제조업체/상표
BRAND 88660
I.D.
3 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
일반 설명
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)