주요 문서
BR37241
BRAND® volumetric flask BLAUBRAND® class A DE-M marking
capacity 10 mL, joint: 7/16, polypropylene stopper
로그인조직 및 계약 가격 보기
크기 선택
2 EA
₩51,461
₩51,461
예상 입고일2025년 5월 30일세부사항
크기 선택
보기 변경
2 EA
₩51,461
About This Item
UNSPSC 코드:
41121804
NACRES:
NB.43
₩51,461
예상 입고일2025년 5월 30일세부사항
material
borosilicate glass 3.3 flask
polypropylene stopper
조인트
joint ST/NS neck7/16
joint: 7/16
특징
class A DE-M marking. DIN EN ISO 1042. Calibrated to contain (TC, In).
포장
pack of 2 ea
제조업체/상표
BRAND 37241
용량
10 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The BRAND® volumetric flask BLAUBRAND® class A with DE-M marking is made from high-quality Boro 3.3 glass and complies with DIN EN ISO 1042 standards. It is calibrated to contain (TC, In), ensuring accurate liquid measurement for various laboratory applications. Each volumetric flask is delivered with a batch number and batch certificate included in the original packaging, providing quality assurance and traceability. This reliable volumetric flask is an essential tool for precise analytical work in any laboratory setting.
법적 정보
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)