These desiccant beads can be regenerated in an oven at 200°C for 3 hours. The product is specially designed for use with Bel-Art Secador® Auto-Desiccator Cabinet, but may also be used as a desiccant in other systems or applications. Please see the link below to review the Secador® Dessicator Cabinet:
https://www.sigmaaldrich.com/substance/sciencewaresecadorautodesiccatorcabinet1234598765
추천 제품
포장
pkg of 26 g
제조업체/상표
Bel-Art F42074-0020
일반 설명
These color-indicating silica gel beads are for use in refilling the Secador® auto-desiccator module which automatically regenerates them for thousands of cycles.
- Silica gel absorbs moisture from the air and is inert, non-toxic and non-corrosive
- Unlike other desiccants, silica gel is non-friable and won’t disintegrate into powder even after repeated regeneration
- Beads contain a color-changing moisture indicator that turns from blue to light pink, indicating that the silica gel is saturated and cannot absorb additional moisture
- When beads turn pink/dark brown, and fail to regain their blue color, it is time to replace the beads with a fresh supply
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
-
I usually dry my beads in an oven. Do these beads require the module to dry them?
1 답변-
도움이 되었습니까?
-