BAF420120000
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
volume 0.09 ft3
동의어(들):
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
로그인조직 및 계약 가격 보기
크기 선택
1 EACH
₩177,625
₩177,625
구입 가능 여부는 고객센터에 문의하십시오.
모든 사진(1)
크기 선택
보기 변경
1 EACH
₩177,625
About This Item
UNSPSC 코드:
41104510
NACRES:
NB.43
₩177,625
구입 가능 여부는 고객센터에 문의하십시오.
추천 제품
material
clear polycarbonate
포장
pkg of 1 ea
제조업체/상표
Bel-Art F42012-0000
용적
0.09 ft3
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
- Lighter and less costly than similarly sized glass models; does not chip like glass
- Holds a full vacuum (29.9" Hg) at room temperature for 24 hours
- Tapered polypropylene stopcock with PTFE plug accepts tubing with ¼ to ⅜” I.D. and allows a controlled entrance of air preventing unnecessary turbulence
- Neoprene O-ring assures a tight seal down to 0 ˚C without the need for grease (ring will not be harmed if grease is used)
- ‘Flat Dome,’ clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
- All models include a 0.32 cm (⅛") thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
- White polypropylene bottom models can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates
- Clear polycarbonate bottom models offer increased viewing visibility from more angles
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
활성 필터