Merck
모든 사진(8)
주요 문서
BAF371110114
SP Bel-Art® Spinbar® magnetic stir bar
cylindrical white PTFE-coated, L × diam. 1.25 in. (31.8 mm) × 0.3125 in. (8 mm)
동의어(들):
Spinbar® magnetic stir bar
로그인조직 및 계약 가격 보기
크기 선택
1 EACH
₩23,853
₩23,853
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
1 EACH
₩23,853
About This Item
UNSPSC 코드:
41122800
NACRES:
NB.51
₩23,853
구입 가능 여부는 고객센터에 문의하십시오.
material
cylindrical white PTFE-coated
포장
pack of 1 ea
제조업체/상표
Bel-Art F37111-0114
길이 × 직경
1.25 in. (31.8 mm) × 0.3125 in. (8 mm)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.
Leakproof, seamless, polished, and contains a permanent Alnico V magnet. Egg-shape bar is designed especially for use in round-bottom vessels.
특징 및 장점
  • Small removable pivot ring in the center adds to their versatility
  • Pivot ring minimizes the contact area of the bar to the vessel, reduces friction and lessens marring of plastic containers
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
법적 정보
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)