주요 문서
BAF249760001
Workshields for β-emitters, free standing
W × L × H 20.3 cm (8 in.) × 30.5 cm (12 in.) × 37.5 cm (14.75 in.), size S
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
26142300
NACRES:
NB.35
크기
S
포장
pack of 1 ea
제조업체/상표
Bel-Art F24976-0001
너비 × 길이 × 높이
20.3 cm (8 in.) × 30.5 cm (12 in.) × 37.5 cm (14.75 in.)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Select shield size to match requirements.
  • 9.5 mm (3/8 in.) thick, glass transparent acrylic shields protect against Beta sources and splashes from lab procedures
  • One piece construction has rounded corner for easy cleaning
  • Non skid feet prevent slipping on benchtop
  • Freestanding design has an angled shield and a flat base with a large work surface
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)