주요 문서
309923
Sigma-Aldrich
Ethyl nitrite solution
10-20 wt. % in ethanol
로그인조직 및 계약 가격 보기
About This Item
Linear Formula:
C2H5ONO
CAS Number:
Molecular Weight:
75.07
MDL number:
UNSPSC 코드:
12352100
PubChem Substance ID:
NACRES:
NA.22
양식
liquid
농도
10-20 wt. % in ethanol
refractive index
n20/D 1.36
density
0.792 g/mL at 25 °C
작용기
O-nitroso
nitroso
저장 온도
2-8°C
SMILES string
CCON=O
InChI
1S/C2H5NO2/c1-2-5-3-4/h2H2,1H3
InChI key
QQZWEECEMNQSTG-UHFFFAOYSA-N
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The reactions of ethyl nitrite solution with human haemoglobin and sperm whale myoglobin were studied[1].
픽토그램
FlameExclamation mark
신호어
Danger
유해 및 위험 성명서
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point (°F)
59.0 °F - closed cup
Flash Point (°C)
15 °C - closed cup
개인 보호 장비
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
이미 열람한 고객
Slide 1 of 1
1 of 1
Takuma Yoshikawa et al.
International journal of pharmaceutics, 583, 119352-119352 (2020-04-24)
Although monoclonal antibodies (mAbs) have revolutionized cancer treatment, their accumulation in solid tumors is limited and requires improvement to enhance therapeutic efficacy. Here we developed a strategy to modify mAb with a donor of nitric oxide (NO) because NO functions
Alessio Baccarani et al.
Journal of reconstructive microsurgery, 23(5), 257-262 (2007-09-19)
Vasospasm leading to thrombosis is a major cause of free flap failure. Flap blood flow may improve by application of vasodilating agents to the pedicle. The purpose of this study is to evaluate the effect of a novel S-nitrosylating agent
Basil M Yurcisin et al.
Annals of surgery, 257(5), 971-977 (2013-01-31)
To determine if reduction in nitric oxide bioactivity contributes to the physiological instability that occurs after brain death and, if so, to also determine in this setting whether administration of a renitrosylating agent could improve systemic physiological status. Organ function
Martin P Moya et al.
Lancet (London, England), 360(9327), 141-143 (2002-07-20)
Inhaled nitric oxide is used to alleviate pulmonary hypertension and hypoxaemia, but generates toxic free radicals and oxides of nitrogen (NO(x)), which can cause rebound-hypoxia and additional pulmonary and other morbidity. To address these problems, we assessed the efficacy of
J C MacDonald et al.
Acta chemica Scandinavica. Series B: Organic chemistry and biochemistry, 35(7), 485-487 (1981-01-01)
Gas chromatography--mass spectrometry demonstrates the presence of ethyl nitrite in the breath of volunteers subjects who smoke and drink concurrently. The mutagenic compound is quantitated at the ppb level by gas chromatography with the selective electron capture detector. The compound