Skip to Content
Merck
All Photos(1)

Key Documents

V001249

Sigma-Aldrich

Titanium(III) chloride solution

15-20%

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
MDL number:
Pricing and availability is not currently available.

Quality Level

concentration

15-20%

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Seidel et al.
The journal of physical chemistry. B, 119(33), 10607-10615 (2015-07-29)
The electronic structure of a Ti(3+) aqueous solution is studied by liquid-jet soft X-ray photoelectron (PE) spectroscopy. Measured valence and Ti 2p core-level binding energies, together with the Ti 2p resonant photoelectron (RPE) spectra and the derived partial electron-yield L-edge
Jeongdae Im et al.
Environmental science & technology, 48(22), 13181-13187 (2014-10-21)
The hydrofluoroolefin 2,3,3,3-tetrafluoropropene (HFO-1234yf) has been introduced to replace 1,1,1,2-tetrafluoroethane (HFC-134a) as refrigerant in mobile, including vehicle, air conditioning systems because of its lower global warming potential. HFO-1234yf is volatile at ambient temperatures; however, high production volumes and widespread handling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service