Skip to Content
Merck
All Photos(1)

Key Documents

V000574

Sigma-Aldrich

Zirconium(IV) oxychloride octahydrate

p.a., 99.5%

Synonym(s):

Zirconium(IV) oxide chloride, Zirconyl chloride octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrOCl2 · 8H2O
CAS Number:
Molecular Weight:
322.25
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

p.a.

Assay

99.5%

SMILES string

O.O.O.O.O.O.O.O.Cl[Zr](Cl)=O

InChI

1S/2ClH.8H2O.O.Zr/h2*1H;8*1H2;;/q;;;;;;;;;;;+2/p-2

InChI key

IZYKRMGFEBSOSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service