Skip to Content
Merck
All Photos(1)

Key Documents

G006021

HPLC Analysis of Peptides on Proteomix® SCX-NP3, Volatile Buffer, ELSD and UV Detection

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777151)
mobile phase: (A) 5 mM ammonium acetate; (B) 50 mM ammonium acetate:acetonitrile (4:1, v:v)
gradient: 0 to 50% B in 20 min
flow rate: 0.6 mL/min
detector: ELSD (SofTA) and UV, 280 nm
injection: 5 μL
sample: 0.1 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service