Skip to Content
Merck
All Photos(1)

Key Documents

33027

Supelco

BSTFA, Derivatization Grade

for GC derivatization

Synonym(s):

N,O-Bis(trimethylsilyl)trifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

for GC derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 25 mL

technique(s)

GC/MS: suitable

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3

InChI key

XCOBLONWWXQEBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

BSTFA is a derivatizing agent widely used in the derivatization of estrogenic steroids, thereby resulting in the formation of trimethylsilyl (TMS) derivative. It is also known to react with the aromatic hydroxyl group.

Application

BSTFA was used for silylation of samples during GC-MS analysis of Glutathione conjugates of Prostaglandin A1.
Suitable for the derivatization of Υ-aminobutyric acid, n-acetylneuraminic acid, arginine, dipeptides, glycine, glycosphingolipi, n-hydroxylamines, lysine, nitrilotriacetate, phenols (sterically hindered), phospholipids, phosphoserine and phosphothreonine, prostaglandins F, prostaglandin H2 methyl ester, prostaglandins F2α, sulfur amino acids, terephthalate prepolymer mixture, thiohydantoins of amino acids, and thromboxane B2.

Other Notes

Reagent for N-acetyl, O-trimethylesilyl, trimethylsilyl, trimethylsilyl diglyceride, trimethylsilyl ether, sulfonyl trimethylsilyl esters and trimethylsilyl thiohydantoins.

related product

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F

Flash Point(C)

34 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N.E. Hoffman and K.A. Peteranetz
Analytical Letters, 5, 589a-589a (1972)
L M Cagen et al.
The Journal of biological chemistry, 251(21), 6550-6554 (1976-11-10)
When prostaglandin A1 was incubated with a Tris/saline suspension of washed human red blood cells, a substantial amount was converted to polar metabolites. These were purified by solvent extraction, XAD-2 column, and cellulose thin layer chromatography and characterized by chromatography
C.W. Gehrke and K. Leimer
Journal of Chromatography A, 57, 219-219 (1971)
A new silylation reagent for amino acids bis(trimethylsilyl)trifluoroacetamide (BSTFA).
D L Stalling et al.
Biochemical and biophysical research communications, 31(4), 616-622 (1968-05-23)
Rapid determination of twenty amino acids by gas chromatography.
J P Hardy et al.
Analytical chemistry, 44(8), 1497-1499 (1972-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service