23395
PTFE Membrane Filter
Zefluor™ membrane, pore size 0.5 μm, O.D. 25 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104900
Recommended Products
material
PTFE (polytetrafluoroethylene)
Zefluor™ membrane
description
for Air Sampling Filter Cassette
feature
with pad
packaging
pkg of 100 ea
O.D.
25 mm
pore size
0.5 μm
application(s)
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
General description
Zefluor™ Membrane Filters are suitable for monitoring acid rain, polynuclear aromatic hydrocarbons (PAHs) and particulates. The membrane filters feature a low-chemical background for highly sensitive determinations. Meets NIOSH specifications.
Legal Information
Zefluor is a trademark of Pall Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service