Z360406
Autoclave pans
capacity 5 L
Synonym(s):
sterilizing pan, sterilizing tray
All Photos(1)
About This Item
Recommended Products
material
polypropylene container
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 6902-5000
L × W × H
324 mm × 257 mm × 108 mm
capacity
5 L
General description
Autoclavable polypropylene with strong rims. Sizes for small instruments and labware. Nestable when empty. Without handles.